missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chlorobenzoic acid, 97%, Thermo Scientific™

Catalog No. AC335970010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC335970010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC335970010 Supplier Thermo Scientific Chemicals Supplier No. 335970010

Chemical Identifiers

CAS 6388-47-2
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00134229
InChI Key LWUAMROXVQLJKA-UHFFFAOYSA-N
Synonym 3-chloroanthranilic acid, benzoic acid, 2-amino-3-chloro, 2-amino-3-chloro-benzoic acid, 3-chloro-2-aminobenzoic acid, anthranilic acid, 3-chloro, 2-amino-3-chlorobenzoicacid, pubchem4639, 3-chloroanthranilicacid, acmc-209nj4, amino-3-chlorobenzoic acid
PubChem CID 80807
IUPAC Name 2-amino-3-chlorobenzoic acid
SMILES NC1=C(Cl)C=CC=C1C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 190°C to 195°C
Color Gray or Tan-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H3ClNH2COOH
Quantity 1 g
Formula Weight 171.58
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-chlorobenzoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-996-4

RUO – Research Use Only