missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-nitrobenzonitrile 98.0+%, TCI America™

Catalog No. p-7117510
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
A15535G 5 g
A155325G 25 g
2 options

Catalog No. A15535G

Supplier: TCI America A15535G

Only null left

Chemical Identifiers

CAS 17601-94-4
Molecular Formula C7H4BrN3O2
Molecular Weight (g/mol) 242.032
MDL Number MFCD00054185
InChI Key MUHLVSZIVTURCZ-UHFFFAOYSA-N
Synonym benzonitrile, 2-amino-3-bromo-5-nitro, 6-bromo-2-cyano-4-nitroaniline, 2-bromo-6-cyano-4-nitroaniline, 2-cyano-4-nitro-6-bromoaniline, 2-amino-3-bromo-5-nitrobenzenecarbonitrile, pubchem2331, 2-amino-3-bromo-1-cyano-5-nitrobenzene, acmc-1bus8, cambridge id 5228140, ksc496g6h
PubChem CID 87173
IUPAC Name 2-amino-3-bromo-5-nitrobenzonitrile
SMILES C1=C(C=C(C(=C1Br)N)C#N)[N+](=O)[O-]

Specifications

Melting Point 191°C
Color Yellow
Quantity 5 g
Formula Weight 242.03
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-bromo-5-nitrobenzonitrile
EINECSNumber (3)-1813
TSCA Yes