missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-bromo-5-methylbenzoic Acid 97.0+%, TCI America™
Supplier: TCI America A25695G
Specifications
| 2-Amino-3-bromo-5-methylbenzoic Acid | |
| 206°C | |
| C8H8BrNO2 | |
| 5 g | |
| LCMZECCEEOQWLQ-UHFFFAOYSA-N | |
| 2-amino-3-bromo-5-methylbenzoic acid | |
| 2774400 | |
| ≥97.0% (T) |
| 13091-43-5 | |
| Yellow | |
| MFCD00051705 | |
| 3-bromo-5-methylanthranilic acid, 2-amino-3-bromo-5-methyl-benzoic acid, 6-amino-5-bromo-m-toluic acid, benzoic acid, 2-amino-3-bromo-5-methyl, acmc-20ao5e, 4-amino-3-bromo-5-carboxytoluene, 2-bromo-6-carboxy-4-methylaniline, benzoic acid,2-amino-3-bromo-5-methyl, 2-azanyl-3-bromanyl-5-methyl-benzoic acid, 2-amino-3-bromo-5-methylbenzoic acid | |
| CC1=CC(Br)=C(N)C(=C1)C(O)=O | |
| 230.06 | |
| 230.06 | |
| Crystalline Powder |
Chemical Identifiers
| 13091-43-5 | |
| 230.06 | |
| LCMZECCEEOQWLQ-UHFFFAOYSA-N | |
| 2-amino-3-bromo-5-methylbenzoic acid |
| C8H8BrNO2 | |
| MFCD00051705 | |
| 2774400 | |
| CC1=CC(Br)=C(N)C(=C1)C(O)=O |
Safety and Handling
TSCA : No