missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-methylbenzoic Acid 97.0+%, TCI America™

Catalog No. A25691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25691G 1 g
A25695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25691G Supplier TCI America Supplier No. A25691G

Chemical Identifiers

CAS 13091-43-5
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD00051705
InChI Key LCMZECCEEOQWLQ-UHFFFAOYSA-N
PubChem CID 2774400
IUPAC Name 2-amino-3-bromo-5-methylbenzoic acid
SMILES CC1=CC(Br)=C(N)C(=C1)C(O)=O

Specifications

Melting Point 206°C
Color Yellow
Quantity 1 g
Synonym 3-bromo-5-methylanthranilic acid, 2-amino-3-bromo-5-methyl-benzoic acid, 6-amino-5-bromo-m-toluic acid, benzoic acid, 2-amino-3-bromo-5-methyl, acmc-20ao5e, 4-amino-3-bromo-5-carboxytoluene, 2-bromo-6-carboxy-4-methylaniline, benzoic acid,2-amino-3-bromo-5-methyl, 2-azanyl-3-bromanyl-5-methyl-benzoic acid, 2-amino-3-bromo-5-methylbenzoic acid
Formula Weight 230.06
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-bromo-5-methylbenzoic Acid