missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5,6-dimethylpyridine 98.0+%, TCI America™

Catalog No. p-7118150
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29251G 1 g
A29255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29251G Supplier TCI America Supplier No. A29251G
Only null left

Chemical Identifiers

CAS 161091-49-2
Molecular Formula C7H9BrN2
Molecular Weight (g/mol) 201.067
MDL Number MFCD04039386
InChI Key PZIWHEZMEJMJLP-UHFFFAOYSA-N
Synonym 6-Amino-5-bromo-2,3-lutidine
PubChem CID 4605855
IUPAC Name 3-bromo-5,6-dimethylpyridin-2-amine
SMILES CC1=CC(=C(N=C1C)N)Br

Specifications

Melting Point 116°C
Quantity 1 g
Formula Weight 201.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-bromo-5,6-dimethylpyridine