Learn More
2-Amino-3,6-dichlorobenzoic acid, 95%, Thermo Scientific Chemicals
Description
2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, acids, bases and sodium hypochlorite.
Specifications
Specifications
| Melting Point | 150°C to 154°C |
| Quantity | 5 g |
| Beilstein | 3266879 |
| Synonym | benzoic acid, 2-amino-3,6-dichloro, 3,6-dichloroanthranilic acid, 2-amino-3,6-dichlorobenzoicacid, acmc-1afqi, 6-amino-2,5-dichlorobenzoic acid, benzoic acid,2-amino-3,6-dichloro, 2-azanyl-3,6-bis chloranyl benzoic acid |
| Solubility Information | Insoluble in water. |
| Formula Weight | 206.03 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-Amino-3,6-dichlorobenzoic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only