missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7044537
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0700306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0700306 Supplier Thermo Scientific Chemicals Supplier No. L0700306

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14438-32-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00017099
InChI Key GIMYRAQQQBFFFJ-UHFFFAOYSA-N
Synonym 3,5-dimethylanthranilic acid, 2-amino-3,5-dimethyl-benzoic acid, 2-amino-3,5-dimethyl benzoic acid, benzoic acid, 2-amino-3,5-dimethyl, pubchem4685, 3,5-dimethylanthranilic, acmc-1bqlm, intermediates-zcf02106, bidd:gt0362, 2-amino-3,5-dimethylbenzoicacid
PubChem CID 259834
IUPAC Name 2-amino-3,5-dimethylbenzoic acid
SMILES CC1=CC(=C(C(=C1)C(=O)O)N)C

Specifications

Melting Point 189°C to 194°C
Quantity 5 g
Beilstein 1101785
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 2-Amino-3,5-dimethylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only