missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dichloropyridine, 97%

Catalog No. AC159381000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159381000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC159381000 Supplier Thermo Scientific Chemicals Supplier No. 159381000
Only null left

CAS: 4214-74-8 | C5H4Cl2N2 | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4214-74-8
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD00006313
InChI Key OCWBGKZFOYMCCN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dichloropyridine, 3,5-dichloro-2-pyridinamine, 2-pyridinamine, 3,5-dichloro, 3,5-dichloro-3-pyridylamine, 3,5-dichloro-2-aminopyridine, 3,5-dichloro-2-pyridylamine, 3,5-dichloropyridin-2-ylamine, pyridine, 2-amino-3,5-dichloro, pubchem13504, acmc-209jnv
PubChem CID 77886
IUPAC Name 3,5-dichloropyridin-2-amine
SMILES NC1=NC=C(Cl)C=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 80°C to 82°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula Cl2(C5H2N)NH2
Quantity 100 g
Beilstein 22, 430
Solubility Information Solubility in water: insoluble
Formula Weight 163.01
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 2-Amino-3, 5-dichloropyridine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if
EINECSNumber 224-143-5
TSCA TSCA

RUO – Research Use Only