accessibility menu, dialog, popup

UserName

NH2NClCl

CAS RN 4214-74-8

IUPAC Name: 3,5-dichloropyridin-2-amine
Synonyms: 3,5-dichloropyridin-2-amine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: OCWBGKZFOYMCCN-UHFFFAOYSA-N
SMILES: NC1=NC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)