missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dichloro-4-methylpyridine 98.0+%, TCI America™

Catalog No. A19321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19321G 1 g
A19325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19321G Supplier TCI America Supplier No. A19321G

Chemical Identifiers

CAS 31430-47-4
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.028
MDL Number MFCD00129031
InChI Key KARIRGBBCOCBGU-UHFFFAOYSA-N
Synonym 2-Amino-3,5-dichloro-4-picoline
PubChem CID 318187
IUPAC Name 3,5-dichloro-4-methylpyridin-2-amine
SMILES CC1=C(C(=NC=C1Cl)N)Cl

Specifications

Melting Point 125°C
Quantity 1 g
Formula Weight 177.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3,5-dichloro-4-methylpyridine