accessibility menu, dialog, popup

UserName

CH3ClNHH2NCl

CAS RN 31430-47-4

IUPAC Name: 2-amino-3,5-dichloro-4-methylpyridin-1-ium
Synonyms: 2-amino-3,5-dichloro-4-methylpyridin-1-ium
Molecular Weight (g/mol): 178.04
Molecular Formula: C6H7Cl2N2
InChi Key: KARIRGBBCOCBGU-UHFFFAOYSA-O
SMILES: CC1=C(Cl)C=[NH+]C(N)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results