missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromopyridine 98.0+%, TCI America™

Catalog No. p-7117536
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15885G 5 g
A158825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15885G Supplier TCI America Supplier No. A15885G

Chemical Identifiers

CAS 35486-42-1
Molecular Formula C5H4Br2N2
Molecular Weight (g/mol) 251.909
MDL Number MFCD00038041
InChI Key WJMJWMSWJSACSN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dibromopyridine, 3,5-dibromo-2-pyridylamine, 2-pyridinamine, 3,5-dibromo, 3,5-dibromo-2-pyridinamine, 3,5-dibromo-pyridin-2-ylamine, 3,5-dibromo-2-pyridinylamine, 2-amino-3,5-dibromo pyridine, 3,5-dibrompyridin-2-amin, pubchem1130, maybridge1_001693
PubChem CID 98851
IUPAC Name 3,5-dibromopyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)Br

Specifications

Melting Point 105°C
Color Yellow
Quantity 5 g
Formula Weight 251.91
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3,5-dibromopyridine