accessibility menu, dialog, popup

UserName

NH2NBrBr

CAS RN 35486-42-1

IUPAC Name: 3,5-dibromopyridin-2-amine
Synonyms: 3,5-dibromopyridin-2-amine
Molecular Weight (g/mol): 251.91
Molecular Formula: C5H4Br2N2
InChi Key: WJMJWMSWJSACSN-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C=C1Br
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)