missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-oxazoline Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117132
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09235G 5 g
A092325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09235G Supplier TCI America Supplier No. A09235G
Only null left

Chemical Identifiers

CAS 375855-07-5
Molecular Formula C3H7ClN2O
Molecular Weight (g/mol) 122.552
MDL Number MFCD00060178
InChI Key HBPBXZZTIWDNKF-UHFFFAOYSA-N
PubChem CID 21475730
IUPAC Name 4,5-dihydro-1,3-oxazol-2-amine;hydrochloride
SMILES C1COC(=N1)N.Cl

Specifications

Melting Point 108°C
Color White
Quantity 5 g
Formula Weight 122.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2-oxazoline Hydrochloride