accessibility menu, dialog, popup

UserName

CAS RN 375855-07-5

ONNH2HCl

CAS RN 375855-07-5

IUPAC Name: 4,5-dihydro-1,3-oxazol-2-amine hydrochloride
Synonyms: 4,5-dihydro-1,3-oxazol-2-amine hydrochloride
Molecular Weight (g/mol): 122.55
Molecular Formula: C3H7ClN2O
InChi Key: HBPBXZZTIWDNKF-UHFFFAOYSA-N
SMILES: Cl.NC1=NCCO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results