missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1-propanol 93.0+%, TCI America™

Catalog No. p-7116842
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A033325ML 25 mL
A0333500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A033325ML Supplier TCI America Supplier No. A033325ML
Only null left

Chemical Identifiers

CAS 124-68-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00008051
InChI Key CBTVGIZVANVGBH-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
PubChem CID 11807
IUPAC Name 2-amino-2-methylpropan-1-ol
SMILES CC(C)(N)CO

Specifications

Color White
Boiling Point 165°C
Quantity 25 mL
Formula Weight 89.14
Percent Purity ≥93.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Amino-2-methyl-1-propanol
EINECSNumber (9)-0118
RTECSNumber UA5950000
TSCA Yes