Learn More
2-Amino-2-methyl-1-propanol, 95%, may cont. ca 5% water
CAS: 124-68-5 | C4H11NO | 89.14 g/mol
Supplier: Thermo Scientific Chemicals A178140F
Description
2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.
Solubility
Immiscible with water. Miscible with alcohols.
Notes
Incompatible withoxidizing agents, strong acids, copper, brass and aluminum.
Specifications
| 2-Amino-2-methyl-1-propanol | |
| 24°C to 28°C | |
| 11.3 | |
| 67°C (152°F) | |
| may cont. ≈5% water | |
| 1.4455 | |
| 2500 mL | |
| 14,449 | |
| Immiscible with water. Miscible with alcohols. | |
| CC(C)(N)CO | |
| 89.14 | |
| 89.14 |
| 124-68-5 | |
| 0.934 | |
| 164°C to 166°C | |
| Amine-like | |
| C4H11NO | |
| MFCD00008051 | |
| 505979 | |
| 2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine | |
| CBTVGIZVANVGBH-UHFFFAOYSA-N | |
| 2-amino-2-methylpropan-1-ol | |
| 11807 | |
| 95% |
Chemical Identifiers
| 124-68-5 | |
| 89.14 | |
| CBTVGIZVANVGBH-UHFFFAOYSA-N | |
| 11807 | |
| CC(C)(N)CO |
| C4H11NO | |
| MFCD00008051 | |
| 2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine | |
| 2-amino-2-methylpropan-1-ol |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P210-P234-P235-P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P370+P378q-P390-P501c
H227-H315-H319-H336-H373
EINECSNumber : 204-709-8
RTECSNumber : UA5950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only