missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1-propanol, 95%, may cont. ca 5% water

Catalog No. AAA17814AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17814AP 500 mL
AAA17814AE 100 mL
AAA178140F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA17814AP Supplier Thermo Scientific Chemicals Supplier No. A17814AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-68-5 | C4H11NO | 89.14 g/mol

2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

Solubility
Immiscible with water. Miscible with alcohols.

Notes
Incompatible withoxidizing agents, strong acids, copper, brass and aluminum.

Chemical Identifiers

CAS 124-68-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00008051
InChI Key CBTVGIZVANVGBH-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
PubChem CID 11807
IUPAC Name 2-amino-2-methylpropan-1-ol
SMILES CC(C)(N)CO

Specifications

Melting Point 24°C to 28°C
Density 0.934
pH 11.3
Boiling Point 164°C to 166°C
Flash Point 67°C (152°F)
Odor Amine-like
Refractive Index 1.4455
Quantity 500 mL
Beilstein 505979
Merck Index 14,449
Solubility Information Immiscible with water. Miscible with alcohols.
Formula Weight 89.14
Percent Purity 95%
Assay may cont. ≈5% water
Chemical Name or Material 2-Amino-2-methyl-1-propanol
Show More Show Less
Hazard Category H227-H315-H319-H336-H373
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P234-P235-P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P370+P378q-P390-P501c
EINECSNumber 204-709-8
RTECSNumber UA5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only