missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-ethyl-1,3-propanediol 98.0+%, TCI America™

Catalog No. p-7116968
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A062025G 25 g
A0620500G 500 g
2 options

Catalog No. A062025G

Supplier: TCI America A062025G

Only null left

Chemical Identifiers

CAS 115-70-8
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.164
MDL Number MFCD00004680
InChI Key IOAOAKDONABGPZ-UHFFFAOYSA-N
Synonym 2-amino-2-ethyl-1,3-propanediol, aepd, aminoethyl propanediol, 1,3-propanediol, 2-amino-2-ethyl, 2-ethyl-2-aminopropanediol, 2-amino-2-ethylpropanediol, unii-0adf1987d4, 2-amino-2-ethyl-propane-1,3-diol, 2-amino-1,3-dihydroxy-2-ethylpropane, aepd-85
PubChem CID 8282
IUPAC Name 2-amino-2-ethylpropane-1,3-diol
SMILES CCC(CO)(CO)N

Specifications

Color White-Yellow
Boiling Point 152°C
Quantity 25 g
Formula Weight 119.16
Percent Purity ≥98.0% (T)
Physical Form Lumps
Chemical Name or Material 2-Amino-2-ethyl-1,3-propanediol
EINECSNumber (2)-0319
TSCA Yes