accessibility menu, dialog, popup

UserName

CH3NH2HOHO

CAS RN 115-70-8

IUPAC Name: 2-amino-2-ethylpropane-1,3-diol
Synonyms: aminoethyl propanediol
Molecular Weight (g/mol): 119.16
Molecular Formula: C5H13NO2
InChi Key: IOAOAKDONABGPZ-UHFFFAOYSA-N
SMILES: CCC(N)(CO)CO
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)