missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-ethyl-1,3-propanediol, 97%

Catalog No. p-7027613
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2450922 100 g
AAB2450936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2450922 Supplier Thermo Scientific Chemicals Supplier No. B2450922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 115-70-8 | C5H13NO2 | 119.164 g/mol

It is used in pharmaceutical industry and in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical industry and in medicine.

Solubility
Fully miscible in water.

Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 115-70-8
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.164
MDL Number MFCD00004680
InChI Key IOAOAKDONABGPZ-UHFFFAOYSA-N
Synonym 2-amino-2-ethyl-1,3-propanediol, aepd, aminoethyl propanediol, 1,3-propanediol, 2-amino-2-ethyl, 2-ethyl-2-aminopropanediol, 2-amino-2-ethylpropanediol, unii-0adf1987d4, 2-amino-2-ethyl-propane-1,3-diol, 2-amino-1,3-dihydroxy-2-ethylpropane, aepd-85
PubChem CID 8282
IUPAC Name 2-amino-2-ethylpropane-1,3-diol
SMILES CCC(CO)(CO)N

Specifications

Density 1.099
Boiling Point 152°C to 153°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.488
Quantity 100 g
Sensitivity Hygroscopic
Merck Index 14,439
Solubility Information Fully miscible in water.
Formula Weight 119.16
Percent Purity 97%
Chemical Name or Material 2-Amino-2-ethyl-1,3-propanediol
Show More Show Less
Hazard Category H318
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 204-101-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only