missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-ethyl-1,3-propanediol, 97%

Catalog No. p-54730
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227115000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227115000 Supplier Thermo Scientific Chemicals Supplier No. 227115000
Only null left

CAS: 115-70-8 | C5H13NO2 | 119.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115-70-8
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00004680
InChI Key IOAOAKDONABGPZ-UHFFFAOYSA-N
Synonym 2-amino-2-ethyl-1,3-propanediol, aepd, aminoethyl propanediol, 1,3-propanediol, 2-amino-2-ethyl, 2-ethyl-2-aminopropanediol, 2-amino-2-ethylpropanediol, unii-0adf1987d4, 2-amino-2-ethyl-propane-1,3-diol, 2-amino-1,3-dihydroxy-2-ethylpropane, aepd-85
PubChem CID 8282
IUPAC Name 2-amino-2-ethylpropane-1,3-diol
SMILES CCC(CO)(CO)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 35°C to 38°C
Color Yellow
Density 1.0990g/mL
Boiling Point 152°C to 153°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4851 to 1.4871
Linear Formula HOCH2C(C2H5)(NH2)CH2OH
Quantity 500 g
Specific Gravity 1.099
Merck Index 15, 436
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 119.16
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 2-Amino-2-ethyl-1, 3-propanediol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
EINECSNumber 204-101-2
TSCA TSCA

RUO – Research Use Only