missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2-Amino-2'-deoxyadenosine, 99%
CAS: 4546-70-7 | C10H14N6O3 | 266.26 g/mol
Supplier: Thermo Scientific Chemicals J6426603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Amino-2'-deoxyadenosine | |
| White | |
| MFCD00047240 | |
| 2-amino-2'-deoxyadenosine, 2,6-diaminopurine-2'-deoxyriboside, adenosine, 2-amino-2'-deoxy, 2,6-diaminopurinedeoxyriboside, 2,6-diaminopurine 2'-deoxyriboside, unii-4zph2f1jhr, 4zph2f1jhr, 9h-purine, 9-2-deoxy-beta-d-erythro-pentofuranosyl-2,6-diamino, 2-amino-deoxyadenosine, 2-amino-2/'-deoxyadenosine | |
| NC1=NC(N)=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=N1 | |
| 266.26 | |
| 266.26 | |
| Powder |
| 4546-70-7 | |
| C10H14N6O3 | |
| 1 g | |
| NOLHIMIFXOBLFF-DWFKHXNSNA-N | |
| (2R,3S,5R)-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol | |
| 97188 | |
| 99% |
Chemical Identifiers
| 4546-70-7 | |
| 266.26 | |
| NOLHIMIFXOBLFF-DWFKHXNSNA-N | |
| 97188 | |
| NC1=NC(N)=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=N1 |
| C10H14N6O3 | |
| MFCD00047240 | |
| 2-amino-2'-deoxyadenosine, 2,6-diaminopurine-2'-deoxyriboside, adenosine, 2-amino-2'-deoxy, 2,6-diaminopurinedeoxyriboside, 2,6-diaminopurine 2'-deoxyriboside, unii-4zph2f1jhr, 4zph2f1jhr, 9h-purine, 9-2-deoxy-beta-d-erythro-pentofuranosyl-2,6-diamino, 2-amino-deoxyadenosine, 2-amino-2/'-deoxyadenosine | |
| (2R,3S,5R)-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol |
Safety and Handling
RTECSNumber : UO7523300
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only