accessibility menu, dialog, popup

UserName

CAS RN 4546-70-7

IUPAC Name: (2R,3S,5R)-5-(2,6-diamino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Synonyms: adenosine, 2-amino-2'-deoxy-
Molecular Weight (g/mol): 266.26
Molecular Formula: C10H14N6O3
InChi Key: NOLHIMIFXOBLFF-DWFKHXNSNA-N
SMILES: NC1=NC(N)=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results