missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Amino-2'-deoxyadenosine, 99%

Catalog No. AAJ6426614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6426614 25 g
AAJ6426603 1 g
AAJ6426606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6426614 Supplier Thermo Scientific Chemicals Supplier No. J6426614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4546-70-7 | C10H14N6O3 | 266.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4546-70-7
Molecular Formula C10H14N6O3
Molecular Weight (g/mol) 266.26
MDL Number MFCD00047240
InChI Key NOLHIMIFXOBLFF-DWFKHXNSNA-N
Synonym 2-amino-2'-deoxyadenosine, 2,6-diaminopurine-2'-deoxyriboside, adenosine, 2-amino-2'-deoxy, 2,6-diaminopurinedeoxyriboside, 2,6-diaminopurine 2'-deoxyriboside, unii-4zph2f1jhr, 4zph2f1jhr, 9h-purine, 9-2-deoxy-beta-d-erythro-pentofuranosyl-2,6-diamino, 2-amino-deoxyadenosine, 2-amino-2/'-deoxyadenosine
PubChem CID 97188
IUPAC Name (2R,3S,5R)-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
SMILES NC1=NC(N)=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=N1

Specifications

Color White
Quantity 25 g
Formula Weight 266.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Amino-2'-deoxyadenosine
RTECSNumber UO7523300
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only