Learn More
Thermo Scientific Chemicals 2'-Amino-2'-deoxyadenosine, 98%
CAS: 10414-81-0 | C10H14N6O3 | 266.26 g/mol
Supplier: Thermo Scientific Chemicals J6553203
Description
2'-AdA can be used as starting material for the synthesis of 2'-amino nucleotides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-AdA can be used as starting material for the synthesis of 2′-amino nucleotides.
Solubility
Soluble in DMSO.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 2'-Amino-2'-deoxyadenosine | |
| White | |
| MFCD06657636 | |
| 2'-amino-2'-deoxyadenosine, 2-amino-adenosine, 2'-deoxy-2'-aminoadenosine, 2r,3s,4r,5r-4-amino-5-6-aminopurin-9-yl-2-hydroxymethyl oxolan-3-ol, 4-amino-5-6-amino-purin-9-yl-2-hydroxymethyl-tetrahydro-furan-3-ol, 2ad, 2-amino-2-deoxyadenosine, adenosine, 2'-amino,-2'-deoxy, 2-nh2-da, adenosine,2'-amino-2'-deoxy | |
| CQKMBZHLOYVGHW-GPGUAWMRNA-N | |
| (2R,3S,4R,5R)-4-amino-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol | |
| 447594 | |
| 98% |
| 10414-81-0 | |
| C10H14N6O3 | |
| 1 g | |
| Soluble in DMSO. | |
| N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=CN=C12 | |
| 266.26 | |
| 266.26 | |
| Powder |
Chemical Identifiers
| 10414-81-0 | |
| 266.26 | |
| CQKMBZHLOYVGHW-GPGUAWMRNA-N | |
| 447594 |
| C10H14N6O3 | |
| MFCD06657636 | |
| 2'-amino-2'-deoxyadenosine, 2-amino-adenosine, 2'-deoxy-2'-aminoadenosine, 2r,3s,4r,5r-4-amino-5-6-aminopurin-9-yl-2-hydroxymethyl oxolan-3-ol, 4-amino-5-6-amino-purin-9-yl-2-hydroxymethyl-tetrahydro-furan-3-ol, 2ad, 2-amino-2-deoxyadenosine, adenosine, 2'-amino,-2'-deoxy, 2-nh2-da, adenosine,2'-amino-2'-deoxy | |
| N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=CN=C12 |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only