missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Amino-2'-deoxyadenosine, 98%

Catalog No. p-7043099
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65532MD 250 mg
AAJ6553203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65532MD Supplier Thermo Scientific Chemicals Supplier No. J65532MD
Only null left

CAS: 10414-81-0 | C10H14N6O3 | 266.26 g/mol

2'-AdA can be used as starting material for the synthesis of 2'-amino nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-AdA can be used as starting material for the synthesis of 2′-amino nucleotides.

Solubility
Soluble in DMSO.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 10414-81-0
Molecular Formula C10H14N6O3
Molecular Weight (g/mol) 266.26
MDL Number MFCD06657636
InChI Key CQKMBZHLOYVGHW-GPGUAWMRNA-N
Synonym 2'-amino-2'-deoxyadenosine, 2-amino-adenosine, 2'-deoxy-2'-aminoadenosine, 2r,3s,4r,5r-4-amino-5-6-aminopurin-9-yl-2-hydroxymethyl oxolan-3-ol, 4-amino-5-6-amino-purin-9-yl-2-hydroxymethyl-tetrahydro-furan-3-ol, 2ad, 2-amino-2-deoxyadenosine, adenosine, 2'-amino,-2'-deoxy, 2-nh2-da, adenosine,2'-amino-2'-deoxy
PubChem CID 447594
SMILES N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=CN=C12

Specifications

Color White
Quantity 250 mg
Solubility Information Soluble in DMSO.
IUPAC Name (2R,3S,4R,5R)-4-amino-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Formula Weight 266.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Amino-2'-deoxyadenosine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only