missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2'-chloro-5-nitrobenzophenone 98.0+%, TCI America™

Catalog No. p-7117584
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A17545G 5 g
A175425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A17545G Supplier TCI America Supplier No. A17545G
Only null left

Chemical Identifiers

CAS 2011-66-7
Molecular Formula C13H9ClN2O3
Molecular Weight (g/mol) 276.68
MDL Number MFCD00792453
InChI Key GRDGBWVSVMLKBV-UHFFFAOYSA-N
Synonym 2-amino-2'-chloro-5-nitrobenzophenone, 2-amino-5-nitrophenyl 2-chlorophenyl methanone, 2-amino-5-nitro-2'-chlorobenzophenone, 2-amino-2'-chloro-5-nitro benzophenone, 2-2-chlorobenzoyl-4-nitroaniline, methanone, 2-amino-5-nitrophenyl 2-chlorophenyl, 2-amino-5-nitrophenyl-2-chlorophenyl methanone, 2-amino-5-nitro-phenyl-2-chloro-phenyl-methanone, 2-amino-5-nitro-2'-chloro benzophenone, pubchem3255
PubChem CID 74830
IUPAC Name 2-(2-chlorobenzoyl)-4-nitroaniline
SMILES NC1=CC=C(C=C1C(=O)C1=CC=CC=C1Cl)[N+]([O-])=O

Specifications

Melting Point 121°C
Color Yellow
Quantity 5 g
Formula Weight 276.68
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2′-chloro-5-nitrobenzophenone