missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-(5-indolyl)acetic acid, 98%

Catalog No. p-7057121
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66482MD 250 mg
AAH6648203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66482MD Supplier Thermo Scientific Chemicals Supplier No. H66482MD

CAS: 108763-43-5 | C10H10N2O2 | 190.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108763-43-5
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD06656882
InChI Key XJJJDZBNTOYJAE-UHFFFAOYSA-N
Synonym 2-amino-2-1h-indol-5-yl acetic acid, amino 1h-indol-5-yl acetic acid, dl-alpha-amino-alpha-indol-5-yl acetic acid, 1h-indole-5-acetic acid, a-amino, 2-amino-2-5-indolyl acetic acid, 1h-indole-5-acetic acid, alpha-amino
PubChem CID 54472597
IUPAC Name 2-amino-2-(1H-indol-5-yl)acetic acid
SMILES C1=CC2=C(C=CN2)C=C1C(C(=O)O)N

Specifications

Color Pink
Quantity 250 mg
Beilstein 15714312
Formula Weight 190.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-2-(5-indolyl)acetic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only