accessibility menu, dialog, popup

UserName

CAS RN 108763-43-5

IUPAC Name: 2-amino-2-(1H-indol-5-yl)acetic acid
Synonyms: amino(1H-indol-5-yl)acetic acid
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: XJJJDZBNTOYJAE-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=CC=C2NC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results