missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-naphthol Hydrochloride 98.0+%, TCI America™

Catalog No. p-7116858
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03651G 1 g
A03655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03651G Supplier TCI America Supplier No. A03651G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41772-23-0
Molecular Formula C10H10ClNO
Molecular Weight (g/mol) 195.646
MDL Number MFCD00050530
InChI Key PEJOQASNBCUDMB-UHFFFAOYSA-N
PubChem CID 12999318
IUPAC Name 2-aminonaphthalen-1-ol;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC(=C2O)N.Cl

Specifications

Quantity 1 g
Formula Weight 195.65
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-1-naphthol Hydrochloride
RTECSNumber QL3340000
TSCA Yes