missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-naphthalenesulfonic Acid 98.0+%, TCI America™

Catalog No. A0343500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0343500G 500 g
A034325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0343500G Supplier TCI America Supplier No. A0343500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81-16-3
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00003988
InChI Key GWIAAIUASRVOIA-UHFFFAOYSA-N
Synonym tobias acid, 2-amino-1-naphthalenesulfonic acid, 2-naphthylamine-1-sulfonic acid, 1-naphthalenesulfonic acid, 2-amino, kyselina tobiasova, unii-f9qkw1fce0, kyselina 2-naftylamin-1-sulfonova, kyselina tobiasova czech, ccris 9048, 2-aminonaphthalene-1-sulphonic acid
PubChem CID 6670
IUPAC Name 2-aminonaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C=CC(=C2S(=O)(=O)O)N

Specifications

Quantity 500 g
Formula Weight 223.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-1-naphthalenesulfonic Acid
EINECSNumber (4)-0493
RTECSNumber QK1250000
TSCA Yes