missing translation for 'onlineSavingsMsg'
Learn More

2-(Allylthio)nicotinic acid, 98%

Catalog No. AAL0709006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0709006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0709006 Supplier Thermo Scientific Chemicals Supplier No. L0709006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 175135-25-8 | C9H9NO2S | 195.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175135-25-8
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00051681
InChI Key KSBMXXMQUGZNSF-UHFFFAOYSA-N
Synonym 2-allylthio nicotinic acid, 2-prop-2-en-1-ylsulfanyl pyridine-3-carboxylic acid, 2-prop-2-enylthiopyridine-3-carboxylic acid, acmc-20aooc, maybridge1_001317, 3-pyridinecarboxylic acid, 2-2-propenylthio, 3-pyridinecarboxylicacid, 2-2-propen-1-ylthio
PubChem CID 1810568
IUPAC Name 2-(prop-2-en-1-ylsulfanyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=C(SCC=C)N=CC=C1

Specifications

Melting Point 144°C to 149°C
Quantity 5 g
Formula Weight 195.24
Percent Purity 98%
Chemical Name or Material 2-(Allylthio)nicotinic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only