accessibility menu, dialog, popup

UserName

H2CSNOOH

CAS RN 175135-25-8

IUPAC Name: 2-(prop-2-en-1-ylsulfanyl)pyridine-3-carboxylic acid
Synonyms: 2-(prop-2-en-1-ylsulfanyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 195.24
Molecular Formula: C9H9NO2S
InChi Key: KSBMXXMQUGZNSF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(SCC=C)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results