missing translation for 'onlineSavingsMsg'
Learn More

2-Allylphenol 98.0+%, TCI America™

Catalog No. p-7116778
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A023325G 25 g
A0233500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A023325G Supplier TCI America Supplier No. A023325G
Only null left

Chemical Identifiers

CAS 1745-81-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00002250
InChI Key QIRNGVVZBINFMX-UHFFFAOYSA-N
Synonym 2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenol
PubChem CID 15624
ChEBI CHEBI:39826
IUPAC Name 2-prop-2-enylphenol
SMILES C=CCC1=CC=CC=C1O

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 g
UN Number 1760
Formula Weight 134.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Allylphenol
EINECSNumber (3)-2840
RTECSNumber SJ3850000
TSCA Yes