missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Allyloxytetrahydropyran 95.0+%, TCI America™
$96.94 - $294.52
Chemical Identifiers
| CAS | 4203-49-0 |
|---|---|
| Molecular Formula | C8H14O2 |
| Molecular Weight (g/mol) | 142.20 |
| MDL Number | MFCD00154089 |
| InChI Key | NRAKYXXDEXCVMV-UHFFFAOYNA-N |
| Synonym | 2-allyloxytetrahydropyran, 2-allyloxy tetrahydro-2h-pyran, 2-allyloxy tetrahydropyran, 2-prop-2-en-1-yloxy oxane, acmc-209jmz, 2-allyloxy-tetrahydro-pyran, allyl tetrahydropyranyl ether, ksc490q3f, tetrahydro-2-allyloxy-2h-pyran |
| PubChem CID | 4532059 |
| IUPAC Name | 2-(prop-2-en-1-yloxy)oxane |
| SMILES | C=CCOC1CCCCO1 |
Chemical Identifiers
| 4203-49-0 | |
| 142.20 | |
| NRAKYXXDEXCVMV-UHFFFAOYNA-N | |
| 4532059 | |
| C=CCOC1CCCCO1 |
| C8H14O2 | |
| MFCD00154089 | |
| 2-allyloxytetrahydropyran, 2-allyloxy tetrahydro-2h-pyran, 2-allyloxy tetrahydropyran, 2-prop-2-en-1-yloxy oxane, acmc-209jmz, 2-allyloxy-tetrahydro-pyran, allyl tetrahydropyranyl ether, ksc490q3f, tetrahydro-2-allyloxy-2h-pyran | |
| 2-(prop-2-en-1-yloxy)oxane |
Specifications
| 4203-49-0 | |
| 75°C | |
| MFCD00154089 | |
| 3271 | |
| NRAKYXXDEXCVMV-UHFFFAOYNA-N | |
| 2-(prop-2-en-1-yloxy)oxane | |
| 4532059 | |
| ≥95.0% (GC) | |
| 2-Allyloxytetrahydropyran |
| Colorless | |
| C8H14O2 | |
| 5 g | |
| 2-allyloxytetrahydropyran, 2-allyloxy tetrahydro-2h-pyran, 2-allyloxy tetrahydropyran, 2-prop-2-en-1-yloxy oxane, acmc-209jmz, 2-allyloxy-tetrahydro-pyran, allyl tetrahydropyranyl ether, ksc490q3f, tetrahydro-2-allyloxy-2h-pyran | |
| C=CCOC1CCCCO1 | |
| 142.20 | |
| 142.20 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator