accessibility menu, dialog, popup

UserName

CAS RN 4203-49-0

H2COO

CAS RN 4203-49-0

IUPAC Name: 2-(prop-2-en-1-yloxy)oxane
Synonyms: 2-(prop-2-en-1-yloxy)oxane
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: NRAKYXXDEXCVMV-UHFFFAOYNA-N
SMILES: C=CCOC1CCCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results