missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-N-Boc-D-glycine dicyclohexylamine salt, 95%

Catalog No. AAH5212703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5212703 1 g
AAH5212706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5212703 Supplier Thermo Scientific Chemicals Supplier No. H5212703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 170899-08-8 | C10H17NO4 | 215.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170899-08-8
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD01321012
InChI Key BUPDPLXLAKNJMI-SSDOTTSWSA-N
Synonym boc-d-allylglycine, r-2-tert-butoxycarbonyl amino pent-4-enoic acid, boc-alpha-allyl-d-gly, boc-d-allyglycine.dcha, 2r-2-tert-butoxycarbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, boc-d-allylglycine.dcha, d-boc-allylglycine, boc-d-allyglycine?dcha, boc-d-allyglycine?cha
PubChem CID 638723
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
SMILES CC(C)(C)OC(=O)NC(CC=C)C(=O)O

Specifications

Quantity 1 g
Formula Weight 396.57
Percent Purity 95%
Chemical Name or Material 2-Allyl-N-Boc-D-glycine dicyclohexylamine salt
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only