missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-4,6-dibenzoylresorcinol, 98%

Catalog No. p-7047333
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1872206 5 g
AAL1872214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1872206 Supplier Thermo Scientific Chemicals Supplier No. L1872206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 102593-74-8 | C23H18O4 | 358.393 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102593-74-8
Molecular Formula C23H18O4
Molecular Weight (g/mol) 358.393
MDL Number MFCD02094038
InChI Key FSYGSBMXRNPJAD-UHFFFAOYSA-N
Synonym 2-allyl-4,6-dibenzoylresorcinol, 5-allyl-4,6-dihydroxy-1,3-phenylene bis phenylmethanone, methanone, 4,6-dihydroxy-5-2-propenyl-1,3-phenylene bis phenyl, 4,6-dibenzoyl-2-prop-2-en-1-yl benzene-1,3-diol, 2allyl-4,6-dibenzoylresorcinol, methanone, 1,1'-4,6-dihydroxy-5-2-propen-1-yl-1,3-phenylene bis 1-phenyl, pubchem21315, acmc-20ao5t
PubChem CID 7010346
IUPAC Name (5-benzoyl-2,4-dihydroxy-3-prop-2-enylphenyl)-phenylmethanone
SMILES C=CCC1=C(C(=CC(=C1O)C(=O)C2=CC=CC=C2)C(=O)C3=CC=CC=C3)O

Specifications

Melting Point 158°C to 163°C
Quantity 5 g
Beilstein 2168154
Formula Weight 358.4
Percent Purity 98%
Chemical Name or Material 2-Allyl-4,6-dibenzoylresorcinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only