missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Allenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™
Supplier: TCI America A20861G
Specifications
| 2-Allenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| Yellow | |
| C9H15BO2 | |
| 1 g | |
| CJAOMXUZZONOSD-UHFFFAOYNA-N | |
| 4,4,5,5-tetramethyl-2-(propa-1,2-dien-1-yl)-1,3,2-dioxaborolane | |
| 11679867 | |
| ≥95.0% (GC,T) |
| 865350-17-0 | |
| 81°C | |
| MFCD09265121 | |
| Allenylboronic Acid Pinacol Ester | |
| CC1(C)OB(OC1(C)C)C=C=C | |
| 166.03 | |
| 166.03 | |
| Liquid |
Chemical Identifiers
| 865350-17-0 | |
| 166.03 | |
| CJAOMXUZZONOSD-UHFFFAOYNA-N | |
| 11679867 | |
| CC1(C)OB(OC1(C)C)C=C=C |
| C9H15BO2 | |
| MFCD09265121 | |
| Allenylboronic Acid Pinacol Ester | |
| 4,4,5,5-tetramethyl-2-(propa-1,2-dien-1-yl)-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator