accessibility menu, dialog, popup

UserName

CAS RN 865350-17-0

BOOH3CCH3H3CH3CCCH2

CAS RN 865350-17-0

IUPAC Name: 4,4,5,5-tetramethyl-2-(propa-1,2-dien-1-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(propa-1,2-dien-1-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 166.03
Molecular Formula: C9H15BO2
InChi Key: CJAOMXUZZONOSD-UHFFFAOYNA-N
SMILES: CC1(C)OB(OC1(C)C)C=C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results