missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Adamantanol 98.0+%, TCI America™
$61.61 - $188.67
Chemical Identifiers
| CAS | 700-57-2 |
|---|---|
| Molecular Formula | C10H16O |
| Molecular Weight (g/mol) | 152.237 |
| MDL Number | MFCD00074744 |
| InChI Key | FOWDOWQYRZXQDP-UHFFFAOYSA-N |
| Synonym | 2-adamantanol, 2-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-2-ol, tricyclo 3.3.1.1∼3,7∼ decan-2-ol, 1r,3r,5r,7r-adamantan-2-ol, 1-adamantanol;, 2alpha-adamantanol, pubchem8753 |
| PubChem CID | 64149 |
| IUPAC Name | adamantan-2-ol |
| SMILES | C1C2CC3CC1CC(C2)C3O |
Chemical Identifiers
| 700-57-2 | |
| 152.237 | |
| FOWDOWQYRZXQDP-UHFFFAOYSA-N | |
| 64149 | |
| C1C2CC3CC1CC(C2)C3O |
| C10H16O | |
| MFCD00074744 | |
| 2-adamantanol, 2-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-2-ol, tricyclo 3.3.1.1∼3,7∼ decan-2-ol, 1r,3r,5r,7r-adamantan-2-ol, 1-adamantanol;, 2alpha-adamantanol, pubchem8753 | |
| adamantan-2-ol |
Specifications
| 700-57-2 | |
| C10H16O | |
| 5 g | |
| FOWDOWQYRZXQDP-UHFFFAOYSA-N | |
| adamantan-2-ol | |
| 64149 | |
| ≥98.0% (GC) | |
| 2-Adamantanol |
| White | |
| MFCD00074744 | |
| 2-adamantanol, 2-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-2-ol, tricyclo 3.3.1.1∼3,7∼ decan-2-ol, 1r,3r,5r,7r-adamantan-2-ol, 1-adamantanol;, 2alpha-adamantanol, pubchem8753 | |
| C1C2CC3CC1CC(C2)C3O | |
| 152.237 | |
| 152.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : No