missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanol, 98%

Catalog No. AAA1069706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1069706 5 g
AAA1069714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1069706 Supplier Thermo Scientific Chemicals Supplier No. A1069706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 700-57-2 | C10H16O | 152.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 700-57-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00074744
InChI Key FOWDOWQYRZXQDP-UHFFFAOYSA-N
Synonym 2-adamantanol, 2-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-2-ol, tricyclo 3.3.1.1∼3,7∼ decan-2-ol, 1r,3r,5r,7r-adamantan-2-ol, 1-adamantanol;, 2alpha-adamantanol, pubchem8753
PubChem CID 64149
IUPAC Name adamantan-2-ol
SMILES C1C2CC3CC1CC(C2)C3O

Specifications

Melting Point 299°C to 303°C (sealed tube)
Quantity 5 g
Beilstein 1903957
Formula Weight 152.24
Percent Purity 98%
Chemical Name or Material 2-Adamantanol
EINECSNumber 211-846-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only