missing translation for 'onlineSavingsMsg'
Learn More

2-Acrylamido-2-methylpropanesulfonic Acid 98.0+%, TCI America™

Catalog No. A092625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A092625G 25 g
A0926100G 100 g
A0926500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A092625G Supplier TCI America Supplier No. A092625G

Chemical Identifiers

CAS 15214-89-8
Molecular Formula C7H13NO4S
Molecular Weight (g/mol) 207.24
MDL Number MFCD00007522
InChI Key XHZPRMZZQOIPDS-UHFFFAOYSA-N
Synonym 2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
PubChem CID 65360
IUPAC Name 2-methyl-2-(prop-2-enamido)propane-1-sulfonic acid
SMILES CC(C)(CS(O)(=O)=O)NC(=O)C=C

Specifications

Melting Point 180°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 207.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acrylamido-2-methylpropanesulfonic Acid
EINECSNumber (2)-2821
TSCA Yes