accessibility menu, dialog, popup

UserName

SNHOOOHOH2CH3CCH3

CAS RN 15214-89-8

IUPAC Name: 2-methyl-2-(prop-2-enamido)propane-1-sulfonic acid
Synonyms: AMPS|2-Methyl-2-[(1-oxo-2-propenyl)amino]-1-propanesulfonic acid
Molecular Weight (g/mol): 207.24
Molecular Formula: C7H13NO4S
InChi Key: XHZPRMZZQOIPDS-UHFFFAOYSA-N
SMILES: CC(C)(CS(O)(=O)=O)NC(=O)C=C
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)