missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acrylamido-2-methylpropanesulfonic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 180°C |
| Color | White |
| Quantity | 100 g |
| UN Number | 3261 |
| Formula Weight | 207.24 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Acrylamido-2-methylpropanesulfonic Acid |
Safety and Handling
| EINECSNumber | (2)-2821 |
| TSCA | Yes |