missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetylpyridine 99.0+%, TCI America™
Supplier: TCI America A0111250ML
Specifications
| 2-Acetylpyridine | |
| Yellow | |
| C7H7NO | |
| 250 mL | |
| AJKVQEKCUACUMD-UHFFFAOYSA-N | |
| 1-pyridin-2-ylethanone | |
| 14286 | |
| ≥99.0% (GC) |
| 1122-62-9 | |
| 189°C | |
| MFCD00006303 | |
| 2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone | |
| CC(=O)C1=CC=CC=N1 | |
| 121.139 | |
| 121.14 | |
| Liquid |
Chemical Identifiers
| 1122-62-9 | |
| 121.139 | |
| AJKVQEKCUACUMD-UHFFFAOYSA-N | |
| 14286 | |
| CC(=O)C1=CC=CC=N1 |
| C7H7NO | |
| MFCD00006303 | |
| 2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone | |
| 1-pyridin-2-ylethanone |
Safety and Handling
EINECSNumber : (5)-3706
RTECSNumber : OB5310000
TSCA : Yes