missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyridine 99.0+%, TCI America™

Catalog No. p-7116721
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A011125ML 25 mL
A0111250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A011125ML Supplier TCI America Supplier No. A011125ML
Only null left

Chemical Identifiers

CAS 1122-62-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00006303
InChI Key AJKVQEKCUACUMD-UHFFFAOYSA-N
Synonym 2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone
PubChem CID 14286
IUPAC Name 1-pyridin-2-ylethanone
SMILES CC(=O)C1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 189°C
Quantity 25 mL
Formula Weight 121.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetylpyridine
EINECSNumber (5)-3706
RTECSNumber OB5310000
TSCA Yes