missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylphenothiazine 98.0+%, TCI America™

Catalog No. p-7118091
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28155G 5 g
A281525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28155G Supplier TCI America Supplier No. A28155G
Only null left

Chemical Identifiers

CAS 6631-94-3
Molecular Formula C14H11NOS
Molecular Weight (g/mol) 241.308
MDL Number MFCD00005017
InChI Key JWGBOHJGWOPYCL-UHFFFAOYSA-N
Synonym 2-acetylphenothiazine, 1-10h-phenothiazin-2-yl ethanone, ethanone, 1-10h-phenothiazin-2-yl, methyl phenothiazin-2-yl ketone, 2-acetyl-10h-phenothiazine, 1-10h-phenothiazin-2-yl ethan-1-one, 3-acetylphenothiazine, 2-acetyl phenothiazine, bidd:gt0700
PubChem CID 81131
IUPAC Name 1-(10H-phenothiazin-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)SC3=CC=CC=C3N2

Specifications

Melting Point 192°C
Quantity 5 g
Formula Weight 241.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetylphenothiazine
TSCA No
Recommended Storage Refrigerator