missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetylnaphtho[2,3-b]furan-4,9-dione 98.0+%, TCI America™
$307.57 - $851.87
Chemical Identifiers
| CAS | 83280-65-3 |
|---|---|
| Molecular Formula | C14H8O4 |
| Molecular Weight (g/mol) | 240.21 |
| MDL Number | MFCD28155270 |
| InChI Key | DPHUWDIXHNQOSY-UHFFFAOYSA-N |
| Synonym | Napabucasin |
| PubChem CID | 10331844 |
| IUPAC Name | 2-acetyl-4H,9H-naphtho[2,3-b]furan-4,9-dione |
| SMILES | CC(=O)C1=CC2=C(O1)C(=O)C1=CC=CC=C1C2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A311025MG
|
TCI America
A311025MG |
25 mg |
Each for $307.57
|
|
|||||
|
A3110100MG
|
TCI America
A3110100MG |
100 mg |
Each for $851.87
|
|
|||||
Chemical Identifiers
| 83280-65-3 | |
| 240.21 | |
| DPHUWDIXHNQOSY-UHFFFAOYSA-N | |
| 10331844 | |
| CC(=O)C1=CC2=C(O1)C(=O)C1=CC=CC=C1C2=O |
| C14H8O4 | |
| MFCD28155270 | |
| Napabucasin | |
| 2-acetyl-4H,9H-naphtho[2,3-b]furan-4,9-dione |
Specifications
| 83280-65-3 | |
| C14H8O4 | |
| 25 mg | |
| DPHUWDIXHNQOSY-UHFFFAOYSA-N | |
| 2-acetyl-4H,9H-naphtho[2,3-b]furan-4,9-dione | |
| 10331844 | |
| ≥98.0% (GC) | |
| 2-Acetylnaphtho[2,3-b]furan-4,9-dione |
| 226°C | |
| MFCD28155270 | |
| Napabucasin | |
| CC(=O)C1=CC2=C(O1)C(=O)C1=CC=CC=C1C2=O | |
| 240.21 | |
| 240.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer